1-(3-Bromophenyl)bicyclo[1.1.1]pentane 95%

Lieferant: Apollo Scientific
Achtung

OR312392-250MG
APOSOR312392-250MGEA 1140 CHF
APOSOR312392-250MG
1-(3-Bromophenyl)bicyclo[1.1.1]pentane 95%
1-(3-Bromophenyl)bicyclo[1.1.1]pentane
Formel: C₁₁H₁₁Br
MW: 223,109 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD27996106
CAS-Nummer: 1823962-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR