tert-Butyl-5-bromoindoline-1-carboxylate

Lieferant: Apollo Scientific

Synonyms: 1-Boc-5-bromoindoline, tert-Butyl 5-bromoindoline-1-carboxylate

OR310451-5G
APOSOR310451-5GEA 151 CHF
APOSOR310451-5G
tert-Butyl-5-bromoindoline-1-carboxylate
tert-Butyl-5-bromoindoline-1-carboxylate
Formel: C₁₃H₁₆BrNO₂
MW: 298.18 g/mol
Melting Pt: 125…127 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08059280
CAS-Nummer: 261732-38-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR