1,3-Diethinylbenzol 97%

Lieferant: Apollo Scientific
Achtung

OR31042-1G OR31042-25G OR31042-5G
APOSOR31042-1GEA 27.9 CHF
APOSOR31042-1G APOSOR31042-25G APOSOR31042-5G
1,3-Diethinylbenzol 97%
1,3-Diethynylbenzene
Formel: C₁₀H₆
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041686
CAS-Nummer: 1785-61-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR