4-Bromo-6-ethoxy-2-methylquinoline

Lieferant: Apollo Scientific

OR309290-1G
APOSOR309290-1GEA 500 CHF
APOSOR309290-1G
4-Bromo-6-ethoxy-2-methylquinoline
4-Bromo-6-ethoxy-2-methylquinoline
Formel: C₁₂H₁₂BrNO
MW: 266.134 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11505131
CAS-Nummer: 1070879-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR