7-Methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309231-1G OR309231-5G
APOSOR309231-1GEA 471 CHF
APOSOR309231-1G APOSOR309231-5G
7-Methylquinoline-3-carboxylic acid
7-Methylquinoline-3-carboxylic acid
Formel: C₁₁H₉NO₂
MW: 187.195 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09787877
CAS-Nummer: 948291-17-6

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR