4-Hydroxy-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-Methyl-1H-indazol-4-ol

OR30842-250MG
APOSOR30842-250MGEA 40.1 CHF
APOSOR30842-250MG
4-Hydroxy-1-methyl-1H-indazole
4-Hydroxy-1-methyl-1H-indazole
Formel: C₈H₈N₂O
MW: 148,162 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11869776
CAS-Nummer: 144528-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR