cis-4-Hydroxy-L(-)-prolin 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline

OR30769-25G
APOSOR30769-25GEA 135 CHF
APOSOR30769-25G
cis-4-Hydroxy-L(-)-prolin 97%
cis-4-Hydroxy-L(-)-prolin
Formel: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 255…257 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064319
CAS-Nummer: 618-27-9
EINECS: 210-542-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR