tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate

Lieferant: Apollo Scientific

Synonyms: tert-Butyl 6-hydroxyindoline-1-carboxylate

OR305478-250MG
APOSOR305478-250MGEA 43.6 CHF
APOSOR305478-250MG
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate
Formel: C₁₃H₁₇NO₃
MW: 235,279 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11045243
CAS-Nummer: 957204-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR