5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific
Achtung

Synonyms: tert-Butyl 5-amino-2,3-dihydro-1H-indole-1-carboxylate, tert-Butyl 5-aminoindoline-1-carboxylate

OR305256-1G
APOSOR305256-1GEA 71.5 CHF
APOSOR305256-1G
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
Formel: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08059270
CAS-Nummer: 129487-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR