2-Methyl-2-propanyl-(3-aminophenyl)carbamat 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic acid tert-Butyl ester, N-Boc-1,3-phenylenediamine, 3-(tert-Butoxycarbonylamino)aniline, N-(tert-Butoxycarbonyl)-1,3-phenylenediamine, tert-Butyl (3-aminophenyl)carbamate, 3-(Boc-amino)aniline

OR305002-100G
APOSOR305002-100GEA 187 CHF
APOSOR305002-100G
2-Methyl-2-propanyl-(3-aminophenyl)carbamat 98%
N-Boc-m-phenylenediamine
Formel: C₁₁H₁₆N₂O₂
MW: 208.261 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06657915
CAS-Nummer: 68621-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR