5-(Cyanomethyl)-2-pyridine carboxylic acid

Lieferant: Apollo Scientific

OR303739-500MG
APOSOR303739-500MGEA 358 CHF
APOSOR303739-500MG
5-(Cyanomethyl)-2-pyridine carboxylic acid
5-(Cyanomethyl)-2-pyridine carboxylic acid
Formel: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 225 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD16140276
CAS-Nummer: 1221791-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR