(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Lieferant: Apollo Scientific

Synonyms: (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, Boc-β-HomoPro-OH

OR302704-5G
APOSOR302704-5GEA 170 CHF
APOSOR302704-5G
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Formel: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01862939
CAS-Nummer: 56502-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR