4-Methoxybenzolcarboximidamidhydrochlorid 97%

Lieferant: Apollo Scientific

Synonyms: 4-Methoxybenzimidamide hydrochloride

OR300727-5G
APOSOR300727-5GEA 107 CHF
APOSOR300727-5G
4-Methoxybenzolcarboximidamidhydrochlorid 97%
4-Methoxybenzolcarboximidamidhydrochlorid
Formel: C₈H₁₀N₂O
MW: 186,64 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00466114
CAS-Nummer: 51721-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR