6-Nitro-o-anisidin

Lieferant: Apollo Scientific

Synonyms: 6-Nitro-o-anisidine, 2-Amino-3-nitroanisole, 2-Methoxy-6-nitrophenylamine, 2-Methoxy-6-nitroaniline

OR300658-1G OR300658-5G
APOSOR300658-1GEA 26.2 CHF
APOSOR300658-1G APOSOR300658-5G
6-Nitro-o-anisidin
6-Nitro-o-anisidin
Formel: C₇H₈N₂O₃
MW: 168.15 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01930197
CAS-Nummer: 16554-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR