6-Chloroimidazo[2,1-b]thiazole-5-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR29800-250MG
APOSOR29800-250MGEA 52.2 CHF
APOSOR29800-250MG
6-Chloroimidazo[2,1-b]thiazole-5-sulphonyl chloride
6-Chloroimidazo[2,1-b]thiazole-5-sulphonyl chloride
Formel: C₅H₂Cl₂N₂O₂S₂
MW: 257,103 g/mol
Melting Pt: 138…140 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00205139
CAS-Nummer: 150020-64-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR