1-(1-Benzofuran-2-yl)ethanon

Lieferant: Apollo Scientific

OR28532-100G
APOSOR28532-100GEA 255 CHF
APOSOR28532-100G
1-(1-Benzofuran-2-yl)ethanon
2-Acetylbenzofuran
Formel: C₁₀H₈O₂
MW: 160.17 g/mol
Siedepunkt: 110…113 °C/3mmHg
Melting Pt: 69…73 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005849
CAS-Nummer: 1646-26-0

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR