2-hydroxy-4,5,6-trimethylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR27042-1G
APOSOR27042-1GEA 410 CHF
APOSOR27042-1G
2-hydroxy-4,5,6-trimethylnicotinonitrile
2-hydroxy-4,5,6-trimethylnicotinonitrile
Formel: C₉H₁₀N₂O
MW: 162,188 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00052650
CAS-Nummer: 91591-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR