2-(1-Ethoxyethoxy)phenol

Lieferant: Apollo Scientific
Achtung

OR26228-100MG
APOSOR26228-100MGEA 168 CHF
APOSOR26228-100MG
2-(1-Ethoxyethoxy)phenol
2-(1-Ethoxyethoxy)phenol
Formel: C₁₀H₁₄O₃
MW: 182.216 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00101502
CAS-Nummer: 51487-88-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR