1,3-Bis(tert-butyl)urea

Lieferant: Apollo Scientific
Achtung

Synonyms: N,N'-Di(tert-butyl)urea, 1,3-Di-tert-butylurea

OR24625-1G
APOSOR24625-1GEA 227 CHF
APOSOR24625-1G
1,3-Bis(tert-butyl)urea
1,3-Bis(tert-butyl)urea
Formel: C₉H₂₀N₂O
MW: 172.268 g/mol
Melting Pt: 238…245 °C
Dichte: 0.974 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00276303
CAS-Nummer: 5336-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR