4-Decylaniline hydrochloride

Lieferant: Apollo Scientific
Achtung

OR22593-10G OR22593-1G OR22593-5G
APOSOR22593-10GEA 415 CHF
APOSOR22593-10G APOSOR22593-1G APOSOR22593-5G
4-Decylaniline hydrochloride
4-Decylaniline hydrochloride
Formel: C₁₆H₂₈ClN
Lager Temperatur: Kühlschrank
MDL Number: MFCD00155303
CAS-Nummer: 175136-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR