Uridin 99%

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

OR18386-100G OR18386-250G OR18386-25G
APOSOR18386-100GEA 95.8 CHF
APOSOR18386-100G APOSOR18386-250G APOSOR18386-25G
Uridin 99%
Uridin
Formel: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 163…169 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00006526
CAS-Nummer: 58-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR