2-(4-Nitrophenyl)ethanol 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol

OR1836-100G OR1836-25G
APOSOR1836-100GEA 208 CHF
APOSOR1836-100G APOSOR1836-25G
2-(4-Nitrophenyl)ethanol 98%
2-(4-Nitrophenyl)ethanol
Formel: C₈H₉NO₃
MW: 167.16 g/mol
Siedepunkt: 177 °C/16mmHg
Melting Pt: 59…62 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010202
CAS-Nummer: 100-27-6
EINECS: 202-835-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR