4-(Cyanomethyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR16985-100MG OR16985-1G OR16985-250MG
APOSOR16985-100MGEA 26.2 CHF
APOSOR16985-100MG APOSOR16985-1G APOSOR16985-250MG
4-(Cyanomethyl)benzoic acid 97%
4-(Cyanomethyl)benzoic acid
Formel: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 196 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00020382
CAS-Nummer: 50685-26-2

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR