2-Chlor-6-nitrobenzothiazol

Lieferant: Apollo Scientific
Achtung

Synonyms: 2-Chloro-6-nitrobenzo[d]thiazole, 2-Chloro-6-nitro-1,3-benzothiazole

OR15153-1G OR15153-5G
APOSOR15153-1GEA 26.2 CHF
APOSOR15153-1G APOSOR15153-5G
2-Chlor-6-nitrobenzothiazol
2-Chloro-6-nitrobenzothiazole
Formel: C₇H₃ClN₂O₂S
Melting Pt: 189…195 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00022852
CAS-Nummer: 2407-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR