1,3-Benzoxazol-5-amin

Lieferant: Apollo Scientific
Achtung

Synonyms: 5-Aminobenzoxazole, 5-Amino-1,3-benzoxazole

OR15063-1G OR15063-5G OR15063-250MG
APOSOR15063-1GEA 105 CHF
APOSOR15063-1G APOSOR15063-5G APOSOR15063-250MG
1,3-Benzoxazol-5-amin
1,3-Benzoxazol-5-amin
Formel: C₇H₆N₂O
Melting Pt: 64…66 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD05256201
CAS-Nummer: 63837-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR