1-Methylindoline-5-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: 5-Formyl-1-methylindoline, 2,3-Dihydro-1-methyl-1H-indole-5-carboxaldehyde

OR14889-50MG
APOSOR14889-50MGEA 26.2 CHF
APOSOR14889-50MG
1-Methylindoline-5-carboxaldehyde 97%
1-Methylindoline-5-carboxaldehyde
Formel: C₁₀H₁₁NO
MW: 161,2 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03208513
CAS-Nummer: 60082-02-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR