1-Brom-4-butylbenzol

Lieferant: Apollo Scientific
Achtung

Synonyms: (4-Bromophenyl)butane

OR1467-100G OR1467-250G OR1467-25G
APOSOR1467-100GEA 76.6 CHF
APOSOR1467-100G APOSOR1467-250G APOSOR1467-25G
1-Brom-4-butylbenzol
1-Brom-4-butylbenzol
Formel: C₁₀H₁₃Br
MW: 213.12 g/mol
Siedepunkt: 242 °C
Melting Pt: -26
Dichte: 1.208 g/cm³ (25 °C)
Flash-Pt: 108 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00040934
CAS-Nummer: 41492-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR