2-Morpholino-5-pyridinboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyms: 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester, 2-(Morpholin-4-yl)pyridine-5-boronic acid pinacol ester, 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine

OR14006-5G
APOSOR14006-5GEA 81.9 CHF
APOSOR14006-5G
2-Morpholino-5-pyridinboronsäurepinakolester
2-Morpholino-5-pyridinboronsäurepinakolester
Formel: C₁₅H₂₃BN₂O₃
MW: 290.17 g/mol
Melting Pt: 132…139 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04039874
CAS-Nummer: 485799-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR