5-Methoxy-1H-indol-3-carbonsäure 98%

Lieferant: Apollo Scientific

Synonyms: 5-Methoxyindole-3-carboxylic acid

OR13726-5G
APOSOR13726-5GEA 135 CHF
APOSOR13726-5G
5-Methoxy-1H-indol-3-carbonsäure 98%
5-Methoxy-1H-indole-3-carboxylic acid
Formel: C10H9NO3
MW: 191.19 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03265451
CAS-Nummer: 10242-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR