5-Chlor-3-methylbenzo[b]thiophen

Lieferant: Apollo Scientific

OR1242-10G
APOSOR1242-10GEA 0 CHF
APOSOR1242-10G
5-Chlor-3-methylbenzo[b]thiophen
5-Chlor-3-methylbenzo[b]thiophen
Formel: C₉H₇ClS
MW: 182,67 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00052506
CAS-Nummer: 19404-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR