2-Amino-6-chlorpyrazin 98%

Lieferant: Apollo Scientific
Achtung

OR11609-25G OR11609-100G OR11609-5G
APOSOR11609-25GEA 61 CHF
APOSOR11609-25G APOSOR11609-100G APOSOR11609-5G
2-Amino-6-chlorpyrazin 98%
2-Amino-6-chloropyrazine
Formel: C₄H₄ClN₃
Melting Pt: 150…152 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00055024
CAS-Nummer: 33332-28-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR