Ethyl-6-hydroxynicotinate 95%

Lieferant: Apollo Scientific

Synonyms: Ethyl 6-hydroxynicotinate

OR110955-25G
APOSOR110955-25GEA 124 CHF
APOSOR110955-25G
Ethyl-6-hydroxynicotinate 95%
Ethyl-6-hydroxynicotinate
Formel: C₈H₉NO₃
MW: 167,1600 g/mol
MDL Number: MFCD08460993
CAS-Nummer: 18617-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR