(2,3-Dihydro-1,4-benzodioxin-6-ylmethylene)malononitrile

Lieferant: Apollo Scientific

OR110854-1G
APOSOR110854-1GEA 0 CHF
APOSOR110854-1G
(2,3-Dihydro-1,4-benzodioxin-6-ylmethylene)malononitrile
(2,3-Dihydro-1,4-benzodioxin-6-ylmethylene)malononitrile
Formel: C₁₂H₈N₂O₂
MW: 212,204 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03619841
CAS-Nummer: 251325-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR