3-Chloroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR110406-1G
APOSOR110406-1GEA 97.5 CHF
APOSOR110406-1G
3-Chloroimidazo[1,2-a]pyridine
3-Chloroimidazo[1,2-a]pyridine
Formel: C₇H₅ClN₂
MW: 152.581 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07778355
CAS-Nummer: 5315-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR