1,8-Naphthyridin-2-carbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR1008-1G
APOSOR1008-1GEA 115 CHF
APOSOR1008-1G
1,8-Naphthyridin-2-carbonsäure 97%
1,8-Naphthyridine-2-carboxylic acid
Formel: C₉H₆N₂O₂
MW: 174.16 g/mol
Melting Pt: 220 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00202900
CAS-Nummer: 215523-34-5

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR