(3,4-Dichlorphenyl)essigsäuremethylester

Lieferant: Apollo Scientific
Achtung

Synonyms: Methyl 3,4-Dichlorophenylacetate, 3,4-Dichlorophenylacetic acid methyll ester, Methyl 2-(3,4-dichlorophenyl)acetate, Methyl (3,4-dichlorophenyl)acetate

OR0617-10G OR0617-25G OR0617-100G
APOSOR0617-10GEA 26.2 CHF
APOSOR0617-10G APOSOR0617-25G APOSOR0617-100G
(3,4-Dichlorphenyl)essigsäuremethylester
(3,4-Dichlorphenyl)essigsäuremethylester
Formel: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219.07 g/mol
Melting Pt: 25…27 °C
Dichte: 1.33 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00800680
CAS-Nummer: 6725-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR