trans-2,6-Dichlor-β-nitrostyrol 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 2,6-Dichloro-β-nitrostyrene

OR0304-1G
APOSOR0304-1GEA 76.6 CHF
APOSOR0304-1G
trans-2,6-Dichlor-β-nitrostyrol 98%
trans-2,6-Dichlor-β-nitrostyrol
Formel: C₈H₅Cl₂NO₂
MW: 218.04 g/mol
Melting Pt: 65 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00024821
CAS-Nummer: 22482-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR