Imidazo[1,2-a]pyrazine 97%

Lieferant: Apollo Scientific
Achtung

OR01539-25G
APOSOR01539-25GEA 168 CHF
APOSOR01539-25G
Imidazo[1,2-a]pyrazine 97%
Imidazo[1,2-a]pyrazine
Formel: C₆H₅N₃
MW: 119.1200 g/mol
Melting Pt: 90…94 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06245371
CAS-Nummer: 274-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR