2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine

Lieferant: Apollo Scientific
Achtung

OR01519-100MG
APOSOR01519-100MGEA 784 CHF
APOSOR01519-100MG
2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine
2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine
Formel: C₁₃H₁₂N₂O
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06245352
CAS-Nummer: 394655-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR