Palladium(II)acetat 99.95+%

Lieferant: Apollo Scientific
Achtung

Synonyms: Palladium diacetate, Palladium (II) acetate

IN3435-1G
APOSIN3435-1GEA 87 CHF
APOSIN3435-1G
Palladium(II)acetat 99.95+%
Palladium(II)acetat
Formel: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 216.3…223.7 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012453
CAS-Nummer: 3375-31-3
EINECS: 222-164-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR