Eisen(II)chlorid Tetrahydrat

Lieferant: Apollo Scientific
Gefahr

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

IN2059-250G IN2059-100G IN2059-1KG IN2059-50G
APOSIN2059-250GEA 131 CHF
APOSIN2059-250G APOSIN2059-100G APOSIN2059-1KG APOSIN2059-50G
Eisen(II)chlorid Tetrahydrat
Eisen(II)chlorid Tetrahydrat
Formel: FeCl₂·4H₂O
MW: 198.81 g/mol
Melting Pt: 105…110 °C
Dichte: 1.93 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149709
CAS-Nummer: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8 II
New Product

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR