Dysprosiumtrinitrat Pentahydrat 99,9%

Lieferant: Apollo Scientific
Gefahr

Synonyms: Dysprosium(III)nitrat Pentahydrat

IN1651-100G IN1651-25G
APOSIN1651-100GEA 599 CHF
APOSIN1651-100G APOSIN1651-25G
Dysprosiumtrinitrat Pentahydrat 99,9%
Dysprosiumtrinitrat Pentahydrat
Formel: DyN₃O₉·5H₂O
Melting Pt: 88.6 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149683
CAS-Nummer: 10031-49-9
EINECS: 233-410-5
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR