Kaliumcarbonat, anhydrous 99%

Lieferant: Apollo Scientific

Synonyms: Potash

IN14572A-100G IN14572A-1KG
APOSIN14572A-100GEA 26.2 CHF
APOSIN14572A-100G APOSIN14572A-1KG
Kaliumcarbonat, anhydrous 99%
Kaliumcarbonat
Formel: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Dichte: 2.43 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011382
CAS-Nummer: 584-08-7
Merck Index: 13,07702

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR