Thymopentin

Lieferant: Apollo Scientific
Achtung

Synonyms: TP-5, (6S,9S,12S,15S,18S)-1,6-Diamino-9-(4-aminobutyl)-12-(carboxymethyl)-18-(4-hydroxybenzyl)-1-imino-15-isopropyl-7,10,13,16-tetraoxo-2,8,11,14,17-pentaazanonadecan-19-oic acid

BITH3057-25MG
APOSBITH3057-25MGEA 69.6 CHF
APOSBITH3057-25MG
Thymopentin
Thymopentin
Formel: C₃₀H₄₉N₉O₉
MW: 679,77 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00214200
CAS-Nummer: 69558-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR