Boceprevir

Lieferant: Apollo Scientific

BISN0326-5MG BISN0326-50MG BISN0326-100MG
APOSBISN0326-5MGEA 112 CHF
APOSBISN0326-5MG APOSBISN0326-50MG APOSBISN0326-100MG
Boceprevir
Boceprevir
Formel: C₂₇H₄₅N₅O₅
MW: 519.68 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD22208555
CAS-Nummer: 394730-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR