Amino-PEG10-alcohol

Lieferant: Apollo Scientific
Achtung

BIPG1051-100MG BIPG1051-250MG BIPG1051-500MG
APOSBIPG1051-100MGEA 184 CHF
APOSBIPG1051-100MG APOSBIPG1051-250MG APOSBIPG1051-500MG
Amino-PEG10-alcohol
Amino-PEG10-alcohol
Formel: C₂₀H₄₃NO₁₀
MW: 457.556 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD28385478
CAS-Nummer: 129449-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR