Biotin-C5-Amine

Lieferant: Apollo Scientific
Achtung

Synonyms: 5-(Biotinamido)pentylamine, N-(5-Aminopentyl)biotinamide, N-(5-Aminopentyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

BIMB102-100MG BIMB102-25MG BIMB102-50MG
APOSBIMB102-100MGEA 0 CHF
APOSBIMB102-100MG APOSBIMB102-25MG APOSBIMB102-50MG
Biotin-C5-Amine
Biotin-C5-Amine
Formel: C₁₅H₂₈N₄O₂S
Melting Pt: 132…135 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00216002
CAS-Nummer: 115416-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR