Okadasäure freie Säure ≥98%

Lieferant: Apollo Scientific
Gefahr

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA

BIM1576-1MG BIM1576-100UG BIM1576-50UG
APOSBIM1576-1MGEA 1610 CHF
APOSBIM1576-1MG APOSBIM1576-100UG APOSBIM1576-50UG
Okadasäure freie Säure ≥98%
Okadaic acid

Potent inhibitor of protein phosphatases, especially the PP-1 and PP-2 classes, in numerous cell types Trends Biochem. Sci. 15: 98-102 (1990).

Formel: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
Melting Pt: 164…166 °C
Dichte: 1.28 g/cm³ (25 °C)
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00083455
CAS-Nummer: 78111-17-8
UN: 3462
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR