Äsculin Sesquihydrat

Lieferant: Apollo Scientific
Achtung

Synonyms: 6,7-Dihydroxycoumarin 6-glucoside sesquihydrate, Esculin sesquihydrate

BIK9005-100G BIK9005-250G BIK9005-50G
APOSBIK9005-100GEA 254 CHF
APOSBIK9005-100G APOSBIK9005-250G APOSBIK9005-50G
Äsculin Sesquihydrat
Äsculin Sesquihydrat
Formel: C₁₅H₁₆O₉·1,5H₂O
MW: 367.31 g/mol
Melting Pt: 195…198 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149492
CAS-Nummer: 66778-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR