Geldanamycin

Lieferant: Apollo Scientific
Achtung

Synonyms: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

BIG2461-100MG BIG2461-250MG BIG2461-25MG
APOSBIG2461-100MGEA 340 CHF
APOSBIG2461-100MG APOSBIG2461-250MG APOSBIG2461-25MG
Geldanamycin
Geldanamycin

Potent antitumour antibiotic.

Formel: C₂₉H₄₀N₂O₉
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00274570
CAS-Nummer: 30562-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR